missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen R/NR3C4 Antibody (AN1-15) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 252.00
Specifications
| Antigen | Androgen R/NR3C4 |
|---|---|
| Clone | AN1-15 |
| Dilution | Western Blot 1:50 - 1:500, Immunohistochemistry 2 - 4 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 2 - 4 μg/mL, Immunohistochemistry-Frozen 2 - 4 μg/mL, Gel Super Shift Assays 1:1 - 1:100, Immunocytochemistry 5 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Androgen R/NR3C4 Monoclonal specifically detects Androgen R/NR3C4 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays.Specifications
| Androgen R/NR3C4 | |
| Western Blot 1:50 - 1:500, Immunohistochemistry 2 - 4 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 2 - 4 μg/mL, Immunohistochemistry-Frozen 2 - 4 μg/mL, Gel Super Shift Assays 1:1 - 1:100, Immunocytochemistry 5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| AR | |
| IgG | |
| Protein A purified | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| AN1-15 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Breast Cancer, Cancer, Cellular Markers | |
| P10275 | |
| 367 | |
| A fusion protein corresponding to residues 331-572 of human androgen receptor. | |
| Primary | |
| Reconstitute with 50 μL sterilized PBS to a final concentration of 1.0 mg/mL. | |
| Androgen receptor. This product does not cross-react with estrogen, progesterone or glucocorticoid receptors. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title