missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen Receptor V7 Antibody (DHTR.V7/9394R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23821-20ug
This item is not returnable.
View return policy
Description
Androgen Receptor V7 Monoclonal antibody specifically detects Androgen Receptor V7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Androgen Receptor V7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| AIS, androgen receptor, AR8, DHTR, dihydrotestosterone receptor, HUMARA, HYSP1, KD, NR3C4, Nuclear receptor subfamily 3 group C member 4, SBMA, SMAX1, TFM | |
| Recombinant fragment (around aa600-920) of human Androgen Receptor-protein (Androgen Receptor-V7 specific) (exact sequence is proprietary) | |
| 20 μg | |
| Breast Cancer, Cancer, Cellular Markers, Phospho Specific | |
| 367 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| DHTR.V7/9394R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction