missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Androgen Receptor V7 Antibody (DHTR.V7/9394R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 268.00 - € 496.00
Specifications
| Antigen | Androgen Receptor V7 |
|---|---|
| Clone | DHTR.V7/9394R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691004
|
Novus Biologicals
NBP3-23821-20ug |
20 μg |
€ 268.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621795
|
Novus Biologicals
NBP3-23821-100ug |
100 μg |
€ 496.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Androgen Receptor V7 Monoclonal antibody specifically detects Androgen Receptor V7 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Androgen Receptor V7 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cellular Markers, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 367 | |
| IgG κ | |
| Protein A purified |
| DHTR.V7/9394R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AIS, androgen receptor, AR8, DHTR, dihydrotestosterone receptor, HUMARA, HYSP1, KD, NR3C4, Nuclear receptor subfamily 3 group C member 4, SBMA, SMAX1, TFM | |
| Recombinant fragment (around aa600-920) of human Androgen Receptor-protein (Androgen Receptor-V7 specific) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title