missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-1 Antibody (MM0019-26B10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB110-85464-20ug
This item is not returnable.
View return policy
Description
Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Angiopoietin-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 20 ug size is provided in liquid form, PBS with No Preservative | |
| AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| No cross reactivity with human Angiopoietin-2 | |
| Human, Chicken | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MM0019-26B10 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:400, Immunohistochemistry-Paraffin 1:50-1:400, Immunohistochemistry-Frozen | |
| Q15389 | |
| ANGPT1 | |
| Human recombinant human Angiopoietin-1 | |
| 20 μg | |
| Angiogenesis, Cancer | |
| 284 | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction