missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-1 Antibody (MM0019-26B10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 232.00 - € 520.00
Specifications
| Antigen | Angiopoietin-1 |
|---|---|
| Clone | MM0019-26B10 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18737603
|
Novus Biologicals
NB110-85464 |
0.1 mg | |||||||
|
18577441
|
Novus Biologicals
NB110-85464-20ug |
20 μg |
€ 232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Angiopoietin-1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer | |
| Q15389 | |
| 284 | |
| Primary | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. |
| MM0019-26B10 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Chicken | |
| AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 | |
| ANGPT1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title