missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiopoietin-1 Antibody (MM0019-26B10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB110-85464
This item is not returnable.
View return policy
Description
Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Angiopoietin-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:400, Immunohistochemistry-Paraffin 1:50-1:400, Immunohistochemistry-Frozen | |
| Q15389 | |
| ANGPT1 | |
| Human recombinant human Angiopoietin-1 | |
| 0.1 mg | |
| Angiogenesis, Cancer | |
| 284 | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MM0019-26B10 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 20 ug size is provided in liquid form, PBS with No Preservative | |
| AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| No cross reactivity with human Angiopoietin-2 | |
| Human, Chicken | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion