missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL7 Monoclonal Antibody (K06358_5H10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556215
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: KHKTPAQPQL KAANCCEEVK ELKAQVANLS SLLSELNKKQ ERDWVSVVMQ VMELESNSKR MESRLTDAES KYSEMNNQID
Angptl7 (angiopoietin-like 7), alternately known as cornea-derived transcript 6 protein (CDT6) or AngX, is a 346 amino acid secreted protein that is expressed specifically in the stromal layer of cornea. Angptl7 forms disulfide-linked homotetramers via coiled-coil interactions and contains one fibrinogen C-terminal domain. Overexpressed in patients with glaucoma, Angptl7 serves as a potential therapeutic target for glaucoma and has been found to increase collagen expression. The gene encoding Angptl7 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.
Specifications
| ANGPTL7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O43827 | |
| ANGPTL7 | |
| Recombinant human ANGPTL7 (immunogen range is 31-110). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K06358_5H10 | |
| Unconjugated | |
| ANGPTL7 | |
| angiopoietin like 7; angiopoietin-like 7; Angiopoietin-like factor; angiopoietin-like factor (CDT6); angiopoietin-like protein 7; Angiopoietin-related protein 7; ANGPTL7; AngX; CDT6; Cornea-derived transcript 6 protein; dJ647M16.1; UNQ313/PRO356 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10218 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction