missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL7 Monoclonal Antibody (K06358_5H10), Invitrogen™
Mouse Monoclonal Antibody
€ 438.00 - € 658.00
Specifications
| Antigen | ANGPTL7 |
|---|---|
| Clone | K06358_5H10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30205740
|
Invitrogen™
MA555835 |
50 μL |
€ 438.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30195753
|
Invitrogen™
MA556215 |
200 μL |
€ 658.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: KHKTPAQPQL KAANCCEEVK ELKAQVANLS SLLSELNKKQ ERDWVSVVMQ VMELESNSKR MESRLTDAES KYSEMNNQID
ANGPTL7 Monoclonal antibody specifically detects ANGPTL7 in Human samples. It is validated for Western BlotSpecifications
| ANGPTL7 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| angiopoietin like 7; angiopoietin-like 7; Angiopoietin-like factor; angiopoietin-like factor (CDT6); angiopoietin-like protein 7; Angiopoietin-related protein 7; ANGPTL7; AngX; CDT6; Cornea-derived transcript 6 protein; dJ647M16.1; UNQ313/PRO356 | |
| Recombinant human ANGPTL7 (immunogen range is 31-110). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ANGPTL7 |
| K06358_5H10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O43827 | |
| 10218 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title