missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD28 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92251-0.1ml
This item is not returnable.
View return policy
Description
ANKRD28 Polyclonal antibody specifically detects ANKRD28 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ANKRD28 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ankyrin repeat domain 28, Ankyrin repeat domain-containing protein 28, KIAA0379PITKPP6-ARS-Aphosphatase interactor targeting K protein, Phosphatase interactor targeting protein hnRNP K, protein phosphatase 6 ankyrin repeat subunit A, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit A, Serine/threonine-protein phosphatase 6 regulatory subunit ARS-A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ANKRD28 (NP_056014.2). MAFLKLRDQPSLVQAIFNGDPDEVRALIFKKEDVNFQDNEKRTPLHAAAYLGDAEIIELLILSGARVNAKDSKWLTPLHRAVASCSEEAVQVLLKHSADVNARDKNWQTPLHIAAANKAVKCAEALVPLLSNVNVSDRAGRTALHHAAFSGHGEMVKLLLSRGANINAFDKKDRRAIHWAAYMGHIEVVKLLVSHGAEVTCKDKKSYTPLHAAASSGMISVVKYLLDLGVDMNEPNAYGNTPLHVACYNGQDVVVNELIDCGAIVNQKNEKGFTPLHFAAASTHGALCLELLVGNGADVN | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| 23243 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction