missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD28 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 463.00
Specifications
| Antigen | ANKRD28 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611421
|
Novus Biologicals
NBP2-92251-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665351
|
Novus Biologicals
NBP2-92251-0.1ml |
0.1 mL |
€ 463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ANKRD28 Polyclonal antibody specifically detects ANKRD28 in Human, Mouse samples. It is validated for Western BlotSpecifications
| ANKRD28 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 23243 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ankyrin repeat domain 28, Ankyrin repeat domain-containing protein 28, KIAA0379PITKPP6-ARS-Aphosphatase interactor targeting K protein, Phosphatase interactor targeting protein hnRNP K, protein phosphatase 6 ankyrin repeat subunit A, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit A, Serine/threonine-protein phosphatase 6 regulatory subunit ARS-A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ANKRD28 (NP_056014.2). MAFLKLRDQPSLVQAIFNGDPDEVRALIFKKEDVNFQDNEKRTPLHAAAYLGDAEIIELLILSGARVNAKDSKWLTPLHRAVASCSEEAVQVLLKHSADVNARDKNWQTPLHIAAANKAVKCAEALVPLLSNVNVSDRAGRTALHHAAFSGHGEMVKLLLSRGANINAFDKKDRRAIHWAAYMGHIEVVKLLVSHGAEVTCKDKKSYTPLHAAASSGMISVVKYLLDLGVDMNEPNAYGNTPLHVACYNGQDVVVNELIDCGAIVNQKNEKGFTPLHFAAASTHGALCLELLVGNGADVN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit