missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ankyrin 3 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05528-50ul
This item is not returnable.
View return policy
Description
Ankyrin 3 Monoclonal antibody specifically detects Ankyrin 3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Ankyrin 3 | |
| Monoclonal | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier | |
| The C-terminal 398 amino acids of human Ankyrin 3 expressed in and purified from E. coli. | |
| 50 μg | |
| Cellular Signaling, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
| 288 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction