Learn More
Ankyrin 3 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
€ 321.00 - € 481.00
Specifications
| Antigen | Ankyrin 3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647597
|
Novus Biologicals
NBP3-05528-100ul |
100 μg |
€ 481.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694069
|
Novus Biologicals
NBP3-05528-50ul |
50 μg |
€ 321.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
Ankyrin 3 Monoclonal antibody specifically detects Ankyrin 3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifikationer
| Ankyrin 3 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier | |
| The C-terminal 398 amino acids of human Ankyrin 3 expressed in and purified from E. coli. | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cellular Signaling, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
| 50% PBS, 50% glycerol | |
| 288 | |
| IgG1 | |
| Immunogen affinity purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel