missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anorthosite from Stillwater Complex, MT, CRM, ARMI∣MBH
Description
Anorthosite from Stillwater Complex, MT, CRM
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Zirconium/Lanthanum/Neodymium/Lead/Gadolinium/Praseodymium/Samarium/Erbium/Molybdenum/Ytterbium/Holmium/Cadmium/Terbium/Lutetium/Manganese/Manganese(II) oxide/Titanium/Titanium dioxide/Potassium/Hafnium/Potassium oxide/Dysprosium/Europium/Magnesium/Rubidium/Iron/Yttrium/Magnesium oxide/Loss on ignition/Total Iron expressed as Fe2O3/Cerium/Sodium/Lithium/Sodium oxide/Scandium/Cobalt/Zinc/Calcium/Calcium oxide/Gallium/Aluminium/Copper/Silicon/Vanadium/Aluminium oxide/Barium/Nickel/Silicon dioxide/Chromium/Strontium |
| CAS | 14808-60-7 |
| Quantity | 75 g |
| Form | Fine Powder |
| Concentration or Composition (by Analyte or Components) | Phosphorus (30) mg/kg/Zirconium (2.3) mg/kg/Lanthanum (0.9) mg/kg/Neodymium (0.7) mg/kg/Lead (0.6) mg/kg/Gadolinium (0.2) mg/kg/Praseodymium (0.2) mg/kg/Samarium (0.2) mg/kg/Erbium (0.1) mg/kg/Molybdenum (0.1) mg/kg/Ytterbium (0.1) mg/kg/Holmium (0.04) mg/kg/Cadmium (0.03) mg/kg/Terbium (0.03) mg/kg/Lutetium (0.02) mg/kg/Manganese 0.0167 %/Manganese(II) oxide 0.0216 %/Titanium 0.035 %/Titanium dioxide 0.058 %/Potassium 0.064 %/Hafnium 0.07 mg/kg/Potassium oxide 0.077 %/Dysprosium 0.206 mg/kg/Europium 0.22 mg/kg/Magnesium 0.78 %/Rubidium 0.93 mg/kg/Iron 1.04 %/Yttrium 1.18 mg/kg/Magnesium oxide 1.29 %/Loss on ignition 1.34 %/Total Iron expressed as Fe2O3 1.48 %/Cerium 1.56 mg/kg/Sodium 1.57 %/Lithium 2 mg/kg/Sodium oxide 2.12 %/Scandium 4.4 mg/kg/Cobalt 6.2 mg/kg/Zinc 9.5 mg/kg/Calcium 11 %/Calcium oxide 15.4 %/Gallium 15.7 mg/kg/Aluminium 16.1 %/Copper 17.4 mg/kg/Silicon 22.2 %/Vanadium 24 mg/kg/Aluminium oxide 30.5 %/Barium 32 mg/kg/Nickel 36 mg/kg/Silicon dioxide 47.4 %/Chromium 56 mg/kg/Strontium 151 mg/kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?