Learn More
CD14 Mouse anti-Human, Alexa Fluor 700, Clone: M5E2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561029
Description
Reacts with the 53-55kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
Host Species: Mouse
Clone: M5E2
Isotype: IgG2a, κ
Cross Reactivity: Human, Rhesus, Cynomolgus, Baboon
Flow Cytometry
Specifications
| CD14 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Human CD14 Protein | |
| 25 μg | |
| Immunology | |
| Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
| IgG2a κ |
| Flow Cytometry | |
| M5E2 | |
| Alexa Fluor 700 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.