missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, Alexa Fluor 700, Clone: M5E2, BD

Mouse Monoclonal Antibody

€ 149.43 - € 270.08

Specifications

Antigen CD14
Clone M5E2
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15870699
View Documents
BD Biosciences
561029
25 μg
ONLINE EXCLUSIVE PRICE
€ 158.00
€ 149.43 / Each
Save € 8.57
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15809118
View Documents
BD Biosciences
557923
0.1 mg
ONLINE EXCLUSIVE PRICE
€ 285.00
€ 270.08 / Each
Save € 14.92
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reacts with the 53-55kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.

Host Species: Mouse
Clone: M5E2
Isotype: IgG2a, κ
Cross Reactivity: Human, Rhesus, Cynomolgus, Baboon

Flow Cytometry

Specifications

Specifications

CD14
0.2mg/mL
Monoclonal
Mouse
Immunology
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
M5E2
Flow Cytometry
Alexa Fluor 700
RUO
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey
Human CD14 Protein
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.