missing translation for 'onlineSavingsMsg'
Learn More

CD90.2 Rat anti-Mouse, Biotin, Clone: 53-2.1, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 553002

 View more versions of this product

Product Code. 15846918

  • ONLINE EXCLUSIVE PRICE
    € 406.00
    € 404.46 / Each
    Save € 1.54
    0.38% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 53-2.1 antibody reacts with the CD90.2 (Thy-1.2) alloantigen on thymocytes, most peripheral T lymphocytes, some intraepithelial T lymphocytes (IEL, DEC), epithelial cells, fibroblasts, neurons, hematopoietic stem cells, but not B lymphocytes, of most mouse strains. mAb 53-2.1 has been reported not to cross-react with Thy-1.1 (e.g., AKR/J, PL), or with rat Thy-1. CD90 is a GPI-anchored membrane glycoprotein of the Ig superfamily which is involved in signal transduction. In addition, there is evidence that CD90 mediates adhesion of
thymocytes to thymic stroma. mAb 53-2.1 has been reported to block the binding of anti-mouse CD90.2 clone 30-H12 (Cat. No. 553009) to immobilized thymocyte membranes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 53-2.1
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse Thymus / Spleen

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD90.2
Monoclonal
0.5mg/mL
Thy-1.2; T25; Thymus cell antigen 1, theta
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
53-2.1
Biotin
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Mouse Thymus / Spleen
0.5 mg
Immunology
Mouse
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.