Learn More
CD90.2 Rat anti-Mouse, Biotin, Clone: 53-2.1, BD
Rat Monoclonal Antibody
€ 404.46
Specifications
| Antigen | CD90.2 |
|---|---|
| Clone | 53-2.1 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
The 53-2.1 antibody reacts with the CD90.2 (Thy-1.2) alloantigen on thymocytes, most peripheral T lymphocytes, some intraepithelial T lymphocytes (IEL, DEC), epithelial cells, fibroblasts, neurons, hematopoietic stem cells, but not B lymphocytes, of most mouse strains. mAb 53-2.1 has been reported not to cross-react with Thy-1.1 (e.g., AKR/J, PL), or with rat Thy-1. CD90 is a GPI-anchored membrane glycoprotein of the Ig superfamily which is involved in signal transduction. In addition, there is evidence that CD90 mediates adhesion of
thymocytes to thymic stroma. mAb 53-2.1 has been reported to block the binding of anti-mouse CD90.2 clone 30-H12 (Cat. No. 553009) to immobilized thymocyte membranes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: 53-2.1
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse Thymus / Spleen
Flow Cytometry
Specifications
| CD90.2 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Immunology | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified | |
| Thy-1.2; T25; Thymus cell antigen 1, theta |
| 53-2.1 | |
| Flow Cytometry | |
| Biotin | |
| RUO | |
| Mouse | |
| Mouse Thymus / Spleen | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.