missing translation for 'onlineSavingsMsg'
Learn More

Ezrin Mouse, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610603

 View more versions of this product

Product Code. 15835539

  • ONLINE EXCLUSIVE PRICE
    € 338.00
    € 329.03 / 1 set
    Save € 8.97
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

First described as an 80kDa protein concentrated in the apical cytoskeletal region of intestinal brush border cells, ezrin is now recognized as a major substrate of protein tyrosine kinases, such as the epidermal growth factor (EGF) tyrosine kinase. Ezrin is expressed at high levels in intestine, kidney, and placenta. In placenta, ezrin is present as monomers and non-covalent oligomers in tight association with actin microfilaments. In the human epidermoid carcinoma cell line A431, microvilli-like structures appear within 30 seconds after the addition of EGF. These structures give way to membrane ruffles after 2-5 minutes, followed by cell rounding after 10-20 minutes. At the same time, ezrin is recruited into these structures and oligomers are formed following its tyrosine phosphorylation. It is thought that tyrosine phosphorylation triggers the formation of ezrin oligomers.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ezrin
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Canine, Human
IgG1
Western Blot
18
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.