missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig κ Light Chain Rat anti-Mouse, PE, Clone: 187.1, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 562021
This item is not returnable.
View return policy
Description
The 187.1 monoclonal antibody specifically binds to kappa light chains of mouse immunoglobulins. The 187.1 antibody does not react with mouse μ1 or μ2 immunoglobulin lights chains or mouse immunoglobulin heavy chains.
Specifications
| Ig κ Light Chain | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2b κ secreted by MPC-11 plasmacytoma | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 187.1 | |
| PE | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction