missing translation for 'onlineSavingsMsg'
Learn More

Ig κ Light Chain Rat anti-Mouse, PE, Clone: 187.1, BD

Rat Monoclonal Antibody

€ 117.30 - € 235.40

Tekniske data

Antigen Ig κ Light Chain
Clone 187.1
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15841569
missing translation for 'viewDocuments'
BD Biosciences
562021
25 μg
ONLINE EXCLUSIVE PRICE
€ 118.00
€ 117.30 / Each
Save € 0.70
0.59% Off
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
15889828
missing translation for 'viewDocuments'
BD Biosciences
559940
0.1 mg
ONLINE EXCLUSIVE PRICE
€ 237.00
€ 235.40 / Each
Save € 1.60
0.68% Off
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 187.1 monoclonal antibody specifically binds to kappa light chains of mouse immunoglobulins. The 187.1 antibody does not react with mouse μ1 or μ2 immunoglobulin lights chains or mouse immunoglobulin heavy chains.

Specifications

Specifications

Ig κ Light Chain
0.2mg/mL
Monoclonal
Rat
Mouse
IgG2b κ secreted by MPC-11 plasmacytoma
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
187.1
Flow Cytometry
PE
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
Videos
SDS
Documents

Documents

Certificates

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.