missing translation for 'onlineSavingsMsg'
Learn More

Ig κ Light Chain Rat anti-Mouse, PE, Clone: 187.1, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 559940

 View more versions of this product

Product Code. 15889828

  • ONLINE EXCLUSIVE PRICE
    € 237.00
    € 235.40 / Each
    Save € 1.60
    0.68% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 187.1 monoclonal antibody specifically binds to kappa light chains of mouse immunoglobulins. The 187.1 antibody does not react with mouse μ1 or μ2 immunoglobulin lights chains or mouse immunoglobulin heavy chains.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ig κ Light Chain
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2b κ secreted by MPC-11 plasmacytoma
0.1 mg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
187.1
PE
Rat
Affinity Purified
RUO
Mouse
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.