Learn More
Anti-Mouse PD-1/CD279 (RMP1-30), Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech 65142-1-IG-500UG
 View more versions of this product
Product Code. 30233162
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The PD-1/CD279 antibody from Proteintech is a rat monoclonal antibody generated with mouse pd-1 transfected bhk cells of mouse PD-1/CD279. This antibody recognizes mouse antigen. The PD-1/CD279 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
PD-1/CD279
Monoclonal
0.5 mg/mL
Rat
Protein A purification
RUO
18566
Store at 2-8°C. Stable for one year after shipment.
Liquid
Flow Cytometry
RMP1-30
Unconjugated
Mouse PD-1 transfected BHK cells
500 μg
Primary
Mouse
Flow Antibody
IgG2b κ