Learn More
p150 [Glued] Mouse, Unlabeled, Clone: 12, BD
Mouse Monoclonal Antibody
€ 249.85
Specifications
| Antigen | p150 [Glued] |
|---|---|
| Clone | 12 |
| Concentration | 250μg/mL |
| Applications | In vitro Assay, Western Blot |
| Classification | Monoclonal |
Description
p150 [Glued] was identified as a polypeptide associated with cytoplasmic dynein,the minus-end-directed microtubule-based motor protein. p150 [Glued] is also a member of the oligomeric dynactin complex. Dynactin mediates dynein-driven vesicle motility, as well as lower eukaryote nuclear transport. p150 [Glued] bears significant homology to the product of the Glued gene in Drosophila. It has been shown in vitro to be a required activator of dynein-mediated transport along microtubules. The p150 [Glued] component of the dynactin complex binds to microtubules and the actin-like protein Centractin (Arp-1), another member of the dynactin complex. In the developing rat, p150 [Glued] is expressed at high levels in neural tissue. Microtubule binding assays with selected constructs of p150 [Glued] indicate that amino acids 39-150 are required for microtubule association.
Immunofluorescence, Western Blotting
Specifications
| p150 [Glued] | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 12 | |
| In vitro Assay, Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human, Mouse, Rat | |
| Rat p150 [Glued] aa. 3-202 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.