Learn More
TCR β Chain Hamster anti-Mouse, APC, Clone: H57-597, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 561080
Description
The H57-597 antibody reacts with a common epitope of the β chain of the T-cell Receptor (TCR) complex on αβ TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β-chain may form homodimers or pair with the pre-TCR α-chain on the surface of immature thymocytes before expression of the TCR α-chain. Plate-bound or soluble H57-597 antibody activates αβ TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
Specifications
| TCR β chain | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| TCR affinity-purified from mouse T-cell hybridoma DO-11.10 | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| H57-597 | |
| APC | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2 λ1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.