missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APC-conjugated IL-6 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech APC-69001-20UL
This item is not returnable.
View return policy
Description
APC-conjugated IL-6 Mouse Monoclonal Antibody, Proteintech shows reactivity in Human samples.Specifications
| IL-6 | |
| Monoclonal | |
| 200 μg/mL | |
| BSF2; HGF; HSF; IFNB2; IL-6 | |
| RecombinantProtein | |
| 20 μL | |
| Primary | |
| Human | |
| Conjugated Antibody | |
| 4 |
| Not Available | |
| 4G3E6 | |
| APC | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3569 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction