Learn More
APOM Rabbit, anti-Human, Clone: 3O19, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 82871-1-RR-20UL
 View more versions of this product
Product Code. 18058175
  • € 149.00 / 20µL
Description
The APOM antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human APOM. This antibody recognizes human antigen. The APOM antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
APOM
3O19
Apo M, apolipoprotein M, APOM, G3a, HSPC336, NG20, Protein G3a
20 μL
55937
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
500 μg/mL
Rabbit
Primary
Human
150 kDa