Learn More
APOM Rabbit, anti-Human, Clone: 3O19, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82871-1-RR-20UL
This item is not returnable.
View return policy
Description
The APOM antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human APOM. This antibody recognizes human antigen. The APOM antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| APOM | |
| 3O19 | |
| Apo M, apolipoprotein M, APOM, G3a, HSPC336, NG20, Protein G3a | |
| 20 μL | |
| 55937 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |