missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOM Rabbit, anti-Human, Clone: 3O19, Proteintech
Rabbit Recombinant Monoclonal Antibody
€ 149.00 - € 439.00
Specifications
| Antigen | APOM |
|---|---|
| Clone | 3O19 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18058175
|
Proteintech
82871-1-RR-20UL |
20 μL |
€ 149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18025475
|
Proteintech
82871-1-RR-100UL |
100 μL |
€ 439.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The APOM antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human APOM. This antibody recognizes human antigen. The APOM antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| APOM | |
| 500 μg/mL | |
| Liquid | |
| Human | |
| Apo M, apolipoprotein M, APOM, G3a, HSPC336, NG20, Protein G3a | |
| Primary |
| 3O19 | |
| Western Blot, ELISA | |
| Rabbit | |
| 55937 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title