missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92417-0.1ml
This item is not returnable.
View return policy
Description
Aquaporin-10 Polyclonal antibody specifically detects Aquaporin-10 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Aquaporin-10 | |
| Polyclonal | |
| Western Blot 1:200-1:1000 | |
| AQP-10, AQPA_HUMAN, Aquaglyceroporin-10, aquaporin 10, aquaporin-10, Small intestine aquaporin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 209-301 of human AQP10 (NP_536354.2). CGIPLNPARDLGPRLFTYVAGWGPEVFSAGNGWWWVPVVAPLVGATVGTATYQLLVALHHPEGPEPAQDLVSAQHKASELETPASAQMLECKL | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 89872 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction