missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARAP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84541
This item is not returnable.
View return policy
Description
ARAP3 Polyclonal specifically detects ARAP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ARAP3 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q8WWN8 | |
| ARAP3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MPNSIYFGLDSRGRAQAAQDKAPDSSQISAPTPALRPTTGTVHIMDPGCLYYGVQPVGTPGAPDRRESRGVCQGRAEHRLSRQDLEAREDAGYASLELPGDSTLLSPTLETEETSDDLISPYASFSF | |
| Affinity Purified | |
| RUO | |
| 64411 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Arf and Rho GAP adapter protein 3, arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 3, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3, ARF-GAP, RHO-GAP, ankyrin repeat and plekstrin homology domains-containingprotein 3, centaurin, delta 3, centaurin-delta-3, CENTD3, Cnt-d3, DRAG1, FLJ21065, phosphoinositide binding protein, PtdIns(34,5)P3-binding protein | |
| Rabbit | |
| 170 kDa | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction