missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARAP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 624.00
Specifications
| Antigen | ARAP3 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18419410
|
Novus Biologicals
NBP1-84541-25ul |
25 μL |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18229507
|
Novus Biologicals
NBP1-84541 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARAP3 Polyclonal specifically detects ARAP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ARAP3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Arf and Rho GAP adapter protein 3, arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 3, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3, ARF-GAP, RHO-GAP, ankyrin repeat and plekstrin homology domains-containingprotein 3, centaurin, delta 3, centaurin-delta-3, CENTD3, Cnt-d3, DRAG1, FLJ21065, phosphoinositide binding protein, PtdIns(34,5)P3-binding protein | |
| ARAP3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8WWN8 | |
| 64411 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MPNSIYFGLDSRGRAQAAQDKAPDSSQISAPTPALRPTTGTVHIMDPGCLYYGVQPVGTPGAPDRRESRGVCQGRAEHRLSRQDLEAREDAGYASLELPGDSTLLSPTLETEETSDDLISPYASFSF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 170 kDa |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto