Learn More
Honeywell™ ARATHERMA NOMEX™ Heavy Gloves
Description
- 100% Nomex™ seamless glove
- Lined with a seamless
- The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
- The glove offers maximum comfort and added sturdiness
- The cotton interlock lining strengthens the glove's mechanical performance while absorbing perspiration
- The glove is ambidextrous and thus more cost-effective
- The glove's relative thickness (gauge 13) enables it to absorb shocks, making it suitable for the handling of heavy objects
- The NOMEX™ fiber pills only a little and ensures good heat resistance for removing dishes from the oven up to 250°C
- White in color, it is compatible with the agrifood industry while enhancing hygiene and cleanliness
Specifications
Specifications
| Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-03 Mechanical Hazards, EN 407-04 Thermical hazards |
| Description | 13 gauge |
| Color | White |
| Hand | Ambidextrous |
| Material | Nomex, Cotton |
| Size | 7 |
| Length (Metric) | 26.5 cm |