missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARC/ARG3.1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92576-0.02ml
This item is not returnable.
View return policy
Description
ARC/ARG3.1 Polyclonal antibody specifically detects ARC/ARG3.1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ARC/ARG3.1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| activity-regulated cytoskeleton-associated protein, ARC/ARG3.1, KIAA0278Arg3.1Activity-regulated gene 3.1 protein homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ARC (NP_056008.1). MELDHRTSGGLHAYPGPRGGQVAKPNVILQIGKCRAEMLEHVRRTHRHLLAEVSKQVERELKGLHRSVGKLESNLDGYVPTSDSQRWKKSIKACLCRCQETIANLERWVKREMHVWREVFYRLERWADRLESTGGKYPVGSESARHTVSVGVGGPESYCHEADGYDYTVSPYAITPPPAAGELPGQEPAEAQQYQPWVPGEDGQPSPGVDTQIFEDPREF | |
| 0.02 mL | |
| Apoptosis, Neuroscience, Sensory Systems | |
| 23237 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction