missing translation for 'onlineSavingsMsg'
Learn More

Arecaidine propargyl ester tosylate, Tocris Bioscience™

Product Code. 18025222
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18025222 10 mg 10mg
18084672 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18025222 Supplier Tocris Bioscience Supplier No. 0383/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Muscarinic agonist

A potent muscarinic receptor agonist. Shows some selectivity for cardiac versus ileal M2 receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147202-94-6
Molecular Formula C17H21NO5S
Molecular Weight (g/mol) 351.417
InChI Key LYHWIOMAWPVTPI-UHFFFAOYSA-N
Synonym arecaidine propargyl ester tosylate, tocris-0383, 4-methylbenzenesulfonic acid; prop-2-ynyl 1-methyl-3,6-dihydro-2h-pyridine-5-carboxylate, n-methyl-1,2,5,6-tetrahydropyridine-3-carboxylic acid propargyl ester tosylate, prop-2-yn-1-yl 1-methyl-5,6-dihydro-2h-pyridine-3-carboxylate; toluenesulfonic acid
PubChem CID 6604936
IUPAC Name 4-methylbenzenesulfonic acid;prop-2-ynyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.CN1CCC=C(C1)C(=O)OCC#C

Specifications

Quantity 10 mg
Formula Weight 351.42
Percent Purity >98%
Chemical Name or Material Arecaidine propargyl ester tosylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.