missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Mouse anti-Bovine, Canine, Chicken, Guinea Pig, Human, Mouse, Porcine, Primate, Rabbit, Rat, Sheep, Clone-ARFS 3F1, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB110-85530-0.025mg
This item is not returnable.
View return policy
Description
ARF1 Monoclonal specifically detects ARF1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ARF1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| P84077 | |
| ARF1 | |
| C-terminal Human ARF1 peptide SNQLRNQ | |
| 0.025 mg | |
| Primary | |
| ARFS 3F1 is Class 1 ARF specific. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| ARFS 3F1 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ADP-ribosylation factor 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 375 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction