missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Mouse anti-Bovine, Canine, Chicken, Guinea Pig, Human, Mouse, Porcine, Primate, Rabbit, Rat, Sheep, Clone-ARFS 3F1, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 190.00
Specifications
| Antigen | ARF1 |
|---|---|
| Clone | ARFS 3F1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot |
Description
ARF1 Monoclonal specifically detects ARF1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| ARF1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| ADP-ribosylation factor 1 | |
| ARF1 | |
| IgG2a | |
| Protein G purified | |
| ARFS 3F1 is Class 1 ARF specific. |
| ARFS 3F1 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Mouse | |
| Human | |
| P84077 | |
| 375 | |
| C-terminal Human ARF1 peptide SNQLRNQ | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title