missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Mouse anti-Human, Mouse, Rabbit, Rat, CoraLite 488, Clone: 1C10E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68069-20UL
This item is not returnable.
View return policy
Description
ARF1 Monoclonal antibody specifically detects ARF1 in Human, Mouse, Rabbit, Rat samples. It is validated for FC (Intra)Specifications
| ARF1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| Mouse | |
| 20 μL | |
| 375 | |
| Store at -20°C | |
| Liquid |
| Flow Cytometry | |
| 1C10E3 | |
| CoraLite 488 | |
| BC011903 | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Rabbit, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction