missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Mouse anti-Human, Mouse, Rabbit, Rat, CoraLite 488, Clone: 1C10E3, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | ARF1 |
|---|---|
| Clone | 1C10E3 |
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17711329
|
Proteintech
CL488-68069-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17701329
|
Proteintech
CL488-68069-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARF1 Monoclonal antibody specifically detects ARF1 in Human, Mouse, Rabbit, Rat samples. It is validated for FC (Intra)Specifications
| ARF1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rabbit, Rat | |
| 375 | |
| Primary | |
| Store at -20°C |
| 1C10E3 | |
| Flow Cytometry | |
| CoraLite 488 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| IgG1 | |
| Protein G purification | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title