missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12541-100ul
This item is not returnable.
View return policy
Description
ARF3 Polyclonal antibody specifically detects ARF3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| ARF3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5ul/mg of lysate | |
| ADP-ribosylation factor 3, small GTP binding protein | |
| Produced in rabbits immunized with purified, recombinant Human ARF3 (Uniprot#: P61204; Met1-Lys181). | |
| 100 μg | |
| Signal Transduction | |
| 377 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction