missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92776-0.1ml
This item is not returnable.
View return policy
Description
ARF6 Polyclonal antibody specifically detects ARF6 in Human, Rat samples. It is validated for Western Blot
Specifications
| ARF6 | |
| Polyclonal | |
| Western Blot 1:50 - 1:200 | |
| ADP-ribosylation factor 6, DKFZp564M0264 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 76-175 of human ARF6 (NP_001654.1). HYYTGTQGLIFVVDCADRDRIDEARQELHRIINDREMRDAIILIFANKQDLPDAMKPHEIQEKLGLTRIRDRNWYVQPSCATSGDGLYEGLTWLTSNYKS | |
| 0.1 mL | |
| Cell Cycle and Replication, Signal Transduction | |
| 382 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction