missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSH Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92083-0.1ml
This item is not returnable.
View return policy
Description
ARSH Polyclonal antibody specifically detects ARSH in Human, Mouse samples. It is validated for Western Blot
Specifications
| ARSH | |
| Polyclonal | |
| Western Blot 1:1000-1:3000 | |
| arylsulfatase family, member H, arylsulfatase H, ASH, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.2, sulfatase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 363-562 of human ARSH (NP_001011719.1). FRWPSVLEAGRVINEPTSLMDIYPTLSYIGGGILSQDRVIDGQNLMPLLEGRASHSDHEFLFHYCGVYLHTVRWHQKDCATVWKAHYVTPKFYPEGTGACYGSGICSCSGDVTYHDPPLLFDISRDPSEALPLNPDNEPLFDSVIKKMEAAIREHRRTLTPVPQQFSVFNTIWKPWLQPCCGTFPFCGCDKEDDILPMAP | |
| 0.1 mL | |
| Cell Biology | |
| 347527 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction