missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSH Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 463.00
Specifications
| Antigen | ARSH |
|---|---|
| Dilution | Western Blot 1:1000-1:3000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18651441
|
Novus Biologicals
NBP2-92083-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615011
|
Novus Biologicals
NBP2-92083-0.1ml |
0.1 mL |
€ 463.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARSH Polyclonal antibody specifically detects ARSH in Human, Mouse samples. It is validated for Western BlotSpecifications
| ARSH | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS with 50% glycerol, pH7.3. | |
| 347527 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:3000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| arylsulfatase family, member H, arylsulfatase H, ASH, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.2, sulfatase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 363-562 of human ARSH (NP_001011719.1). FRWPSVLEAGRVINEPTSLMDIYPTLSYIGGGILSQDRVIDGQNLMPLLEGRASHSDHEFLFHYCGVYLHTVRWHQKDCATVWKAHYVTPKFYPEGTGACYGSGICSCSGDVTYHDPPLLFDISRDPSEALPLNPDNEPLFDSVIKKMEAAIREHRRTLTPVPQQFSVFNTIWKPWLQPCCGTFPFCGCDKEDDILPMAP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title