Learn More
Invitrogen™ ASPM Monoclonal Antibody (K52002_11G4), Invitrogen™
Mouse Monoclonal Antibody
€ 480.00 - € 846.00
Specifications
| Antigen | ASPM |
|---|---|
| Clone | K52002_11G4 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30203775
|
Invitrogen™
MA556234 |
200 μg |
€ 846.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30196946
|
Invitrogen™
MA555854 |
50 μg |
€ 480.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: CKVPLSNENS QVPQSPEDWR KSEVSPRIPE CQGSKSPKAI FEELVEMKSN YYSFIKQNNP KFSAVQDISS HSHNKQPKRR PILSATVTKR KATCTRENQT EINKPKAKRC LNSAVGEHEK VINNQKEKED FHSYLPIIDP ILSKSKSYKN EVTPSSTTAS VARKRKSDGS MEDANVRVAI TEHTEVREIK RIHFSPSEPK TSAVKKTKNV TTPISKRISN REKLNLKKKT DLSIFRTPIS KTNKRTKPII AVAQSSLTFI KPLKTDIPRH PMPFAAKNMF YDERWKEKQE QGFTWWLNFI LTPDDFTVKT
ASPM Monoclonal antibody specifically detects ASPM in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ASPM | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| abnormal spindle microtubule assembly; Abnormal spindle protein homolog; Abnormal spindle-like microcephaly-associated protein; ASP; asp (abnormal spindle) homolog, microcephaly associated; Asp homolog; ASPM; Calmbp1; MCPH5 | |
| Recombinant human ASPM (immunogen range is 411-720). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ASPM |
| K52002_11G4 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8IZT6 | |
| 259266 | |
| IgG2b | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.