missing translation for 'onlineSavingsMsg'
Learn More

Asporin Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications

Brand:  Novus Biologicals NB100-1514

 View more versions of this product

Product Code. 18798273

  • € 544.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Asporin Polyclonal specifically detects Asporin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Asporin
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
asporin, asporin (LRR class 1), FLJ20129, OS3, periodontal ligament associated protein 1, Periodontal ligament-associated protein 1, PLAP-1, PLAP1SLRR1Casporin proteoglycan, small leucine-rich protein 1C
Goat
43 kDa
0.1 mg
Primary
Expected from sequence similarity: Rat.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
0.5 mg/mL
Western Blot 0.1-1 ug/ml, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 10 ug/ml, Peptide ELISA detection limit 1:64000, Immunocytochemistry
Q9BXN1
ASPN
Peptide with sequence C-IHENKVKKIQKDT corresponding to internal region according to NP_060150.3.
Affinity Purified
RUO
54829
Human, Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.