missing translation for 'onlineSavingsMsg'
Learn More

Asporin Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications

€ 247.00 - € 544.00

Specifications

Antigen Asporin
Concentration 0.5 mg/mL
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18798273
View Documents
Novus Biologicals
NB100-1514
0.1 mg
€ 544.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18584313
View Documents
Novus Biologicals
NB100-1514-25ug
25 μg
€ 247.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Asporin Polyclonal specifically detects Asporin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications

Specifications

Asporin
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
asporin, asporin (LRR class 1), FLJ20129, OS3, periodontal ligament associated protein 1, Periodontal ligament-associated protein 1, PLAP-1, PLAP1SLRR1Casporin proteoglycan, small leucine-rich protein 1C
ASPN
IgG
Affinity Purified
0.5 mg/mL
Polyclonal
Goat
Human, Mouse, Rat
Q9BXN1
54829
Peptide with sequence C-IHENKVKKIQKDT corresponding to internal region according to NP_060150.3.
Primary
43 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.