missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asporin Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-1514-25ug
This item is not returnable.
View return policy
Description
Asporin Polyclonal specifically detects Asporin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| Asporin | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| asporin, asporin (LRR class 1), FLJ20129, OS3, periodontal ligament associated protein 1, Periodontal ligament-associated protein 1, PLAP-1, PLAP1SLRR1Casporin proteoglycan, small leucine-rich protein 1C | |
| Goat | |
| 43 kDa | |
| 25 μg | |
| Primary | |
| Expected from sequence similarity: Rat. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 0.1-1 μg/mL, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL, Peptide ELISA detection limit 1:64000, Immunocytochemistry | |
| Q9BXN1 | |
| ASPN | |
| Peptide with sequence C-IHENKVKKIQKDT corresponding to internal region according to NP_060150.3. | |
| Affinity Purified | |
| RUO | |
| 54829 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction