Learn More
Karl Hecht™ Assistent™ Glass Vials
Product Code. p-8290028
Click to view available options
Volume (Metric):
8 mL
20 mL
Height (Metric):
34 mm
60 mm
Diameter (Metric) Outer:
24 mm
26 mm
Unit Size:
Pack of 130
Pack of 154
Product Code. 10145291
Brand: Karl Hecht™ 42783012
Only null left
Add to basket
Add to basket
Suitable for media, diagnostic, storage, display and sample collection applications. Karl Hecht™ Assistent™ Glass Vials come with a plastic snap-cap for secure closure.
  • Clear glass
  • With rolled rim
Specifications
Diameter (Metric) Outer 24 mm
Volume (Metric) 8 mL
Closure Type Snap Cap
Height (Metric) 34 mm