Learn More
ATG5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 211 publications
Brand: Novus Biologicals NB110-53818
This item is not returnable.
View return policy
Description
ATG5 Polyclonal specifically detects ATG5 in Human, Mouse, Rat, Porcine, Alligator, Bovine, Drosophila, Fish, Guinea Pig, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Radioimmunoassay, Proximity Ligation Assay, Electron Microscopy, Knockout Validated, Knockdown Validated.
Specifications
| ATG5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| APG5 autophagy 5-like (S. cerevisiae), ASPAPG5-LIKE, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is selective for the full-length and calpain cleaved isoform proteins. The short isoform is missing amino acids 1-79. The calpain cleaved form of ATG5 is missing amino acids 195-275. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, Radioimmune Assays (RIA), In situ PLA, Immunoassay, KnockDown | |
| 1.0 mg/ml | |
| Western Blot 1:500, Simple Western 1:50, Flow Cytometry, ELISA, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:400, Immunoblotting, Radioimmunoassay, Proximity Ligation Assay, Electron Microscopy, Knockout Validated, KnockDown Validated | |
| Q9H1Y0 | |
| ATG5 | |
| A synthetic peptide made to an N-terminal region of the human ATG5 protein (within residues 1-50) [Swiss-Prot Q9H1Y0] | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Cell Biology, Ubiquitin Proteasome Pathway | |
| 9474 | |
| Human, Mouse, Rat, Pig, Alligator, Bovine, Drosophila, Fish, Guinea Pig, Primate, Xenopus species, Zebrafish | |
| IgG |