missing translation for 'onlineSavingsMsg'
Learn More

ATG5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 211 publications

€ 315.00 - € 625.00

Specifications

Antigen ATG5
Concentration 1.0 mg/ml
Dilution Western Blot 1:500, Simple Western 1:50, Flow Cytometry, ELISA, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:400, Immunoblotting, Radioimmunoassay, Proximity Ligation Assay, Electron Microscopy, Knockout Validated, KnockDown Validated
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, Radioimmune Assays (RIA), In situ PLA, Immunoassay, KnockDown
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18786403
View Documents
Novus Biologicals
NB110-53818SS
0.025 mL
€ 315.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18735813
View Documents
Novus Biologicals
NB110-53818
0.1 mL
€ 625.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ATG5 Polyclonal specifically detects ATG5 in Human, Mouse, Rat, Porcine, Alligator, Bovine, Drosophila, Fish, Guinea Pig, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Radioimmunoassay, Proximity Ligation Assay, Electron Microscopy, Knockout Validated, Knockdown Validated.
Specifications

Specifications

ATG5
Western Blot 1:500, Simple Western 1:50, Flow Cytometry, ELISA, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:400, Immunoblotting, Radioimmunoassay, Proximity Ligation Assay, Electron Microscopy, Knockout Validated, KnockDown Validated
Polyclonal
Rabbit
Apoptosis, Autophagy, Cancer, Cell Biology, Ubiquitin Proteasome Pathway
PBS with 0.02% Sodium Azide
APG5 autophagy 5-like (S. cerevisiae), ASPAPG5-LIKE, ATG5 autophagy related 5 homolog (S. cerevisiae), Autophagy protein 5
ATG5
IgG
Affinity Purified
This is selective for the full-length and calpain cleaved isoform proteins. The short isoform is missing amino acids 1-79. The calpain cleaved form of ATG5 is missing amino acids 195-275.
1.0 mg/ml
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, Radioimmune Assays (RIA), In situ PLA, Immunoassay, KnockDown
Unconjugated
RUO
Human, Mouse, Rat, Pig, Alligator, Bovine, Drosophila, Fish, Guinea Pig, Primate, Xenopus species, Zebrafish
Q9H1Y0
9474
A synthetic peptide made to an N-terminal region of the human ATG5 protein (within residues 1-50) [Swiss-Prot Q9H1Y0]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.