missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG7 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81760-1-RR-20UL
This item is not returnable.
View return policy
Description
The ATG7 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human ATG7. This antibody recognizes human antigen. The ATG7 antibody has been validated for the following applications: WB, IF/ICC, IP, ELISA analysis.Specifications
| ATG7 | |
| Recombinant Monoclonal | |
| 500 μg/mL | |
| APG7-LIKE; APG7L; DKFZp434N0735; GSA7 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 68-78 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| 2M8 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 10533 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction