missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG7 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
€ 149.00 - € 439.00
Specifications
| Antigen | ATG7 |
|---|---|
| Clone | 2M8 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233510
|
Proteintech
81760-1-RR-20UL |
20 μL |
€ 149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236207
|
Proteintech
81760-1-RR-100UL |
100 μL |
€ 439.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ATG7 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human ATG7. This antibody recognizes human antigen. The ATG7 antibody has been validated for the following applications: WB, IF/ICC, IP, ELISA analysis.Specifications
| ATG7 | |
| 500 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 10533 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2M8 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| APG7-LIKE; APG7L; DKFZp434N0735; GSA7 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title